Skip to Content
Merck
All Photos(1)

Key Documents

47524

Sigma-Aldrich

Fmoc-tBu-Gly-OH

≥98.0%

Synonym(s):

Fmoc-L-tert-leucine, Fmoc-Tle-OH

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€291.00

€291.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€291.00

About This Item

Empirical Formula (Hill Notation):
C21H23NO4
CAS Number:
Molecular Weight:
353.41
Beilstein:
6662856
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

€291.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98.0%

form

solid

reaction suitability

reaction type: C-H Activation
reaction type: Fmoc solid-phase peptide synthesis
reagent type: ligand
reaction type: Peptide Synthesis

mp

124-127 °C

application(s)

peptide synthesis

functional group

Fmoc
amine
carboxylic acid

storage temp.

2-8°C

SMILES string

CC(C)(C)[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C21H23NO4/c1-21(2,3)18(19(23)24)22-20(25)26-12-17-15-10-6-4-8-13(15)14-9-5-7-11-16(14)17/h4-11,17-18H,12H2,1-3H3,(H,22,25)(H,23,24)/t18-/m1/s1

InChI key

VZOHGJIGTNUNNC-GOSISDBHSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service