Skip to Content
Merck
All Photos(1)

Key Documents

474932

Sigma-Aldrich

2,5-Dichlorophenyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NCS
CAS Number:
Molecular Weight:
204.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.67 (lit.)

bp

145-147 °C/15 mmHg (lit.)

density

1.425 g/mL at 25 °C (lit.)

SMILES string

Clc1ccc(Cl)c(c1)N=C=S

InChI

1S/C7H3Cl2NS/c8-5-1-2-6(9)7(3-5)10-4-11/h1-3H

InChI key

JHTPBGFVWWSHDL-UHFFFAOYSA-N

General description

2,5-Dichlorophenyl isothiocyanate, an isothiocyanate derivative, is also referred to as 1,4-dichloro-2-isothiocyanatobenzene. Its enthalpy of vaporization at boiling point (536.82K) has been reported to be 47.433kjoule/mol.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 535-535 (2014)
Fazila Rizvi et al.
Scientific reports, 9(1), 6738-6738 (2019-05-03)
A library of thiosemicarbazide derivatives of isoniazid 3-27, was synthesized and evaluated for their anti-inflammatory and urease inhibition activities, by using in vitro bioassays. Among these compounds 9, 10, 12, 21, and 26 were identified as new derivatives. Prolonged use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service