Skip to Content
Merck
All Photos(1)

Key Documents

473707

Sigma-Aldrich

4,4-Dimethoxy-2-methyl-2-butanol

97%

Synonym(s):

β-Hydroxyisovaleraldehyde dimethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2CHCH2C(CH3)2OH
CAS Number:
Molecular Weight:
148.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.4226 (lit.)

density

0.956 g/mL at 25 °C (lit.)

SMILES string

COC(CC(C)(C)O)OC

InChI

1S/C7H16O3/c1-7(2,8)5-6(9-3)10-4/h6,8H,5H2,1-4H3

InChI key

RLJXZLBECCIWMU-UHFFFAOYSA-N

General description

4,4-Dimethoxy-2-methyl-2-butanol is an organic building block.

Application

4,4-Dimethoxy-2-methyl-2-butanol may be used in the microwave-mediated synthesis of 2,2-dimethyl-2H-chromones.[1] It may also be used in the preparation of alkyl substituted 3′R,4′R-di-O-(-)-camphanoyl-2′,2′-dimethyldihydropyrano[2,3-f]chromone (DCP) analogs.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Zhou et al.
Bioorganic & medicinal chemistry, 18(18), 6678-6689 (2010-08-24)
In a continued study, 23 3'R,4'R-di-O-(-)-camphanoyl-2',2'-dimethyldihydropyrano[2,3-f]chromone (DCP) derivatives (5-27) were synthesized, and screened for anti-HIV activity against both a non-drug-resistant NL4-3 strain and multiple reverse transcriptase (RT) inhibitor-resistant (RTMDR-1) strain, using 2-EDCP (4) and 2-MDCP (35) as controls. New DCP
Ting Zhou et al.
Tetrahedron letters, 51(33), 4382-4386 (2010-10-12)
A novel and efficient microwave-assisted one-pot reaction was developed to synthesize angular 2,2-dimethyl-2H-chromone containing compounds, which is the first and key step in the synthesis of potent DCK and DCP anti-HIV agents. The newly developed microwave synthesis conditions dramatically shortened

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service