Skip to Content
Merck
All Photos(1)

Documents

472751

Sigma-Aldrich

6-O-(tert-Butyldimethylsilyl)-D-glucal

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24O4Si
CAS Number:
Molecular Weight:
260.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D +90°, c = 1 in chloroform

refractive index

n20/D 1.485 (lit.)

density

1.06 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)[Si](C)(C)OC[C@H]1OC=C[C@@H](O)[C@@H]1O

Application

Important building block for both solution- and solid-phase synthesis of oligosaccharides.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seeberger, P.H., et al.
Aldrichimica Acta, 30, 75-75 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service