Skip to Content
Merck
All Photos(1)

Key Documents

471941

Sigma-Aldrich

Potassium tetrakis(2-thienyl)borate

Synonym(s):

Tetra-2-thienylborate potassium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12BKS4
CAS Number:
Molecular Weight:
382.44
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

reaction suitability

core: boron
reagent type: catalyst

mp

>350 °C (lit.)

SMILES string

[K+].c1csc(c1)[B-](c2cccs2)(c3cccs3)c4cccs4

InChI

1S/C16H12BS4.K/c1-5-13(18-9-1)17(14-6-2-10-19-14,15-7-3-11-20-15)16-8-4-12-21-16;/h1-12H;/q-1;+1

InChI key

HNYWOQMQXVVVRO-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Dienes, trienes, and cyclohexenes by coupling of organic halides, internal alkynes or cyclohexadienes and organoboranes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service