Skip to Content
Merck
All Photos(1)

Key Documents

470708

Sigma-Aldrich

Methyl 3-hydroxy-4-methoxybenzoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CO2CH3
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

64-67 °C (lit.)

SMILES string

COC(=O)c1ccc(OC)c(O)c1

InChI

1S/C9H10O4/c1-12-8-4-3-6(5-7(8)10)9(11)13-2/h3-5,10H,1-2H3

InChI key

QXOXUEFXRSIYSW-UHFFFAOYSA-N

Application

Methyl 3-hydroxy-4-methoxybenzoate may be used as starting reagent in the novel synthesis of gefitinib. Synthesis involves the alkylation of starting material, followed by nitration, reduction, cyclization, chlorination and two successive amination reactions. It may be used as starting reagent in the synthesis of methyl 5-(benzo[d][1,3]dioxol-5-yl)-4-methoxy-2-nitrobenzoate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew S Rosenthal et al.
Bioorganic & medicinal chemistry letters, 21(10), 3152-3158 (2011-04-01)
Continued examination of substituted 6-arylquinazolin-4-amines as Clk4 inhibitors resulted in selective inhibitors of Clk1, Clk4, Dyrk1A and Dyrk1B. Several of the most potent inhibitors were validated as being highly selective within a comprehensive kinome scan.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service