Skip to Content
Merck
All Photos(2)

Key Documents

469068

Sigma-Aldrich

1-Bromobenzocyclobutene

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€221.00

€221.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€221.00

About This Item

Empirical Formula (Hill Notation):
C8H7Br
CAS Number:
Molecular Weight:
183.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€221.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

refractive index

n20/D 1.59 (lit.)

bp

90 °C/10.5 mmHg (lit.)

density

1.45 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

BrC1Cc2ccccc12

InChI

1S/C8H7Br/c9-8-5-6-3-1-2-4-7(6)8/h1-4,8H,5H2

InChI key

AYNXHFRDABNHRX-UHFFFAOYSA-N

General description

1-Bromobenzocyclobutene is a useful synthon. It has important applications in organometallic methodology.[1] Reaction between cycloheptatriene, bromoform, potassium carbonate and 18-crown-6 at 140°C yields 1-bromobenzocyclobutene.[2]

Application

1-Bromobenzocyclobutene may be used in the synthesis of following compounds:
  • five-membered zirconacycles[1]
  • benzocyclobutenol and benzocyclobutenone[3]
  • 2,3-dimethoxyprotoberberinium bromide, via reaction with 3,4-dihydro-6,7-dimethoxyisoquinoline[4]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies on the syntheses of heterocyclic compounds-DXLV: An alternative synthesis of the protoberberine ring system.
Kametani T, et al.
Tetrahedron, 30(9), 1043-1046 (1974)
1-Bromobenzocyclobutene: a convenient entry into the benzocyclobutene ring system.
DeCamp MR and Viscogliosi LA.
The Journal of Organic Chemistry, 46(19), 3918-3920 (1981)
Condensed Cyclobutane Aromatic Compounds. IX. Benzocyclobutenol and Benzocyclobutenone.
Cava MP and Muth K.
Journal of the American Chemical Society, 82(3), 652-654 (1960)
T V V Ramakrishna et al.
Organic letters, 5(6), 877-879 (2003-03-14)
[reaction: see text] Commercially available 1-bromobenzocyclobutene is a potentially useful synthon particularly with the application of organometallic methodology. Here we show that it is readily converted into Cp(2)Zr(benzocyclobutadiene), which couples with alkynes or nitriles giving five-membered zirconacycles. Treatment of these

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service