Skip to Content
Merck
All Photos(2)

Key Documents

468975

Sigma-Aldrich

Benzyl (S)-(−)-4-oxo-2-azetidinecarboxylate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€270.00

€270.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€270.00

About This Item

Empirical Formula (Hill Notation):
C11H11NO3
CAS Number:
Molecular Weight:
205.21
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€270.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

optical activity

[α]21/D −44°, c = 3.3 in chloroform

mp

137-140 °C (lit.)

functional group

ester
phenyl

SMILES string

O=C1C[C@H](N1)C(=O)OCc2ccccc2

InChI

1S/C11H11NO3/c13-10-6-9(12-10)11(14)15-7-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,12,13)/t9-/m0/s1

InChI key

WGLLBHSIXLWVFU-VIFPVBQESA-N

Application

Benzyl (S)-(-)-4-oxo-2-azetidinecarboxylate may be used in the preparation of its N-acetyl derivative, which can be a potential candidate as solid electrolyte interphase (SEI)-forming additive in lithium-ion batteries.[1]
Important building block for the synthesis of NMDA receptor antagonists,[2] 3-alkyl-L-aspartic acids,[3] and orally active β-lactam inhibitors.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Computational screening of lactam molecules as solid electrolyte interphase forming additives in lithium-ion batteries.
Han YK, et al.
Current Applied Physics, 14(6), 897-900 (2014)
Hanessian, S. et al.
Synlett, 33-33 (1992)
Baldwin, J.E. et al.
Tetrahedron, 51, 11581-11581 (1995)
P E Finke et al.
Journal of medicinal chemistry, 38(13), 2449-2462 (1995-06-23)
The stereospecific synthesis of several 4-[(4-carboxyphenyl)oxy]- 3,3-dialkyl-1-[[(1-phenylalkyl)-amino]carbonyl]azetidin-2-on es 3 is described in which the C-3 alkyl groups were varied from methyl to butyl as well as allyl, benzyl and methoxymethyl. The structure-activity relations for these compounds are discussed in terms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service