468452
Dipotassium tris(1,2-benzenediolato-O,O′)silicate
97%
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
K2Si(O2C6H4)3
CAS Number:
Molecular Weight:
430.57
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
97%
form
solid
reaction suitability
core: potassium
mp
360 °C (lit.)
SMILES string
[K+].[K+].O1c2ccccc2O[Si--]1345Oc6ccccc6O3.O4c7ccccc7O5
InChI
1S/C18H12O6Si.2K/c1-2-8-14-13(7-1)19-25(20-14,21-15-9-3-4-10-16(15)22-25)23-17-11-5-6-12-18(17)24-25;;/h1-12H;;/q-2;2*+1
InChI key
NRFBMPOQXYVYMM-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Silica's versatility spans various industries, including biomedical applications.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service