468118
Poly(tetrafluoroethylene)
powder, 100 μm particle size
Synonym(s):
Fluorinated polymer, Fluoropolymer, PTFE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
powder
particle size
100 μm
transition temp
Tm 321 °C
density
2.1 g/mL at 25 °C
SMILES string
F\C(F)=C(\F)F
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of thoracic and cardiovascular surgery, 150(1), 152-157 (2015-03-15)
Robotically assisted cardiac surgery is an alternative to conventional, open-chest surgery. Although studies have been done on the clinical effect, morbidity, and mortality of robotically assisted atrial myxoma excision, few have addressed surgical outcomes, such as pain, quality of life
PLoS biology, 13(8), e1002213-e1002213 (2015-08-14)
Repair of DNA breaks by single-strand annealing (SSA) is a major mechanism for the maintenance of genomic integrity. SSA is promoted by proteins (single-strand-annealing proteins [SSAPs]), such as eukaryotic RAD52 and λ phage Redβ. These proteins use a short single-stranded
Scientific reports, 5, 8419-8419 (2015-02-13)
Stretchable electronics offers unsurpassed mechanical compliance on complex or soft surfaces like the human skin and organs. To fully exploit this great advantage, an autonomous system with a self-powered energy source has been sought for. Here, we present a new
Anesthesiology, 122(1), 117-126 (2014-08-30)
Multicapillary column ion-mobility spectrometry (MCC-IMS) may identify volatile components in exhaled gas. The authors therefore used MCC-IMS to evaluate exhaled gas in a rat model of sepsis, inflammation, and hemorrhagic shock. Male Sprague-Dawley rats were anesthetized and ventilated via tracheostomy
Nature communications, 5, 5254-5254 (2014-10-17)
Despite recent progress in preparing numerous types of nanosheets, it remains a difficult challenge to assemble the tiny building blocks into functional macroscale architectures suitable for practical applications. Here we introduce a diffusion driven layer-by-layer assembly process and demonstrate its
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service