Skip to Content
Merck
All Photos(1)

Key Documents

467693

Sigma-Aldrich

4-Fluorothioanisole

97%

Synonym(s):

4-Fluorophenyl methyl sulfide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€66.60

€66.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€66.60

About This Item

Linear Formula:
FC6H4SCH3
CAS Number:
Molecular Weight:
142.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€66.60


Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.551 (lit.)

bp

184-185 °C (lit.)

density

1.167 g/mL at 25 °C (lit.)

functional group

fluoro
thioether

SMILES string

CSc1ccc(F)cc1

InChI

1S/C7H7FS/c1-9-7-4-2-6(8)3-5-7/h2-5H,1H3

InChI key

XFUMHENRNCUHOH-UHFFFAOYSA-N

General description

4-Fluorothioanisole is thioanisole derivative. Its conformations about the Csp2-S bond have been investigated by J method.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Motion about the Csp 2-S bond in thioanisole and some derivatives by the J method.
Schaefer T and Baleja JD.
Canadian Journal of Chemistry, 64(7), 1326-1331 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service