Skip to Content
Merck
All Photos(1)

Key Documents

467278

Sigma-Aldrich

N-Methyl-o-toluidine

≥95%

Synonym(s):

2,N-Dimethylaniline, 2-Methyl-N-methylaniline, N,2-Dimethylaniline, N,2-Dimethylbenzenamine, N,o-Dimethylaniline, N-(2-Methylphenyl)methylamine, N-Methyl-2-methylaniline, N-Methyl-2-methylbenzenamine, N-Methyl-N-(o-tolyl)amine, N-Methyl-o-methylaniline, o,N-Dimethylaniline, Methyl(o-tolyl)amine, NSC 9395

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NHCH3
CAS Number:
Molecular Weight:
121.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

bp

177-204 °C/740 mmHg (lit.)

density

0.981 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CNc1ccccc1C

InChI

1S/C8H11N/c1-7-5-3-4-6-8(7)9-2/h3-6,9H,1-2H3

InChI key

GUAWMXYQZKVRCW-UHFFFAOYSA-N

General description

N-Methyl-o-toluidine can be synthesized by reacting o-toluidine and benzotriazole.

Application

N-Methyl-o-toluidine may be used in the preparation of 2-substituted N-methylindoles.
Dye intermediate
Reactant for:
  • Palladium-catalyzed coupling reactions
  • Palladium-catalyzed amination of bromoindazoles

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

174.9 °F - closed cup

Flash Point(C)

79.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaakko Laakia et al.
Talanta, 132, 889-893 (2014-12-06)
Eight selected isomeric amines were ionized using atmospheric pressure chemical ionization and atmospheric pressure photoionization producing a protonated molecule [M+H](+) for each amine. The mobility of these ions was measured by ion mobility spectrometry. The amine compound class was shown
Carbon dioxide: a reagent for the simultaneous protection of nucleophilic centers and the activation of alternative locations to electrophilic attack. XVI, A novel synthetic method for the side-chain functionalization of N-methyl-o-toluidine and for the preparation of 2-substituted N-methylindoles.
Katritzky AR, et al.
Heterocycles, 30(1), 407-414 (1990)
A Practical Synthetic Method for N-Methyl-o-Toluidine.
Katritzky AR and Akutagawa K.
Organic preparations and procedures international, 21(3), 340-341 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service