Skip to Content
Merck
All Photos(2)

Key Documents

466700

Sigma-Aldrich

Tri-O-benzoyl-D-galactal

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€194.00

€194.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€194.00

About This Item

Empirical Formula (Hill Notation):
C27H22O7
CAS Number:
Molecular Weight:
458.46
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

€194.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

SMILES string

O=C(OC[C@H]1OC=C[C@@H](OC(=O)c2ccccc2)[C@H]1OC(=O)c3ccccc3)c4ccccc4

InChI

1S/C27H22O7/c28-25(19-10-4-1-5-11-19)32-18-23-24(34-27(30)21-14-8-3-9-15-21)22(16-17-31-23)33-26(29)20-12-6-2-7-13-20/h1-17,22-24H,18H2/t22-,23-,24-/m1/s1

InChI key

OZFFEFRJEYIEGH-WXFUMESZSA-N

Application

Important building block for both solution- and solid-phase synthesis of oligosaccharides.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seeberger, P.H., et al.
Aldrichimica Acta, 30, 75-75 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service