Skip to Content
Merck
All Photos(1)

Key Documents

464295

Sigma-Aldrich

N,N-Dimethylnonylamine

97%

Synonym(s):

1-(Dimethylamino)nonane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8N(CH3)2
CAS Number:
Molecular Weight:
171.32
Beilstein:
1736553
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.428 (lit.)

bp

83 °C/7 mmHg (lit.)

density

0.773 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCCCCCCCN(C)C

InChI

1S/C11H25N/c1-4-5-6-7-8-9-10-11-12(2)3/h4-11H2,1-3H3

InChI key

AMAADDMFZSZCNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylnonylamine (1-(Dimethylamino)nonane) is a hydrophobic tertiary amine.

Application

N,N-Dimethylnonylamine may be used as charge modifier for the separation of enantiomers of anti-inflammatory drugs. It may be used for the synthesis of bromoethylnonyldimethylammonium bromide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ouyang et al.
Bioconjugate chemistry, 11(1), 104-112 (2000-01-19)
A series of novel cationic detergents that contain cleavable hydrophilic isothiuronium headgroups was synthesized, and their utility in controlled assembly of plasmid DNA into small stable particles with high DNA concentration investigated. The detergents have alkyl chains of C(8)-C(12) and
Dynamic modification of the chiral bonding properties of a CHIRAL-AGP column by organic and inorganic additives: separation of enantiomers of anti-inflammatory drugs.
Hermansson J and Hermansson I.
Journal of Chromatography A, 666(1), 181-191 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service