463051
Diborane
99.99% (diborane only), 9-11% (balance hydrogen), 10% in hydrogen, electronic grade
Synonym(s):
Borane (B2 H6 ), Boron hydride (B2 H6 ), Diboron hexahydride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
B2H6
CAS Number:
Molecular Weight:
27.67
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
grade
electronic grade
Assay
99.99% (diborane only)
form
gas
reaction suitability
reagent type: reductant
concentration
10% in hydrogen
9-11% (balance hydrogen)
bp
−92.5 °C (lit.)
SMILES string
BB
InChI
1S/B2H4/c1-2/h1-2H2
InChI key
QSJRRLWJRLPVID-UHFFFAOYSA-N
General description
Atomic number of base material: 5 Boron
Application
Dibrorane gas is used primarily as a p-type dopant in the deposition of eptaxial and amorphous silicon thin films.
Useful as a p-type dopant for the deposition of silicon-containing thin films.
Packaging
Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.
Compatible with the following:
Compatible with the following:
- Aldrich® lecture-bottle station systems
- Aldrich® lecture-bottle gas regulators
Other Notes
See Technical Information Bulletin AL-151 Gas Regulators: Selection, Installation, and Operation
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
also commonly purchased with this product
Product No.
Description
Pricing
control valve
hose barb
Product No.
Description
Pricing
purge valve
Product No.
Description
Pricing
recommended
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Flam. Gas 1
Storage Class Code
2A - Gases
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Miquel Torrent-Sucarrat et al.
The journal of physical chemistry. A, 109(4), 615-621 (2006-07-13)
In previous works we have shown that certain pi-conjugated organic molecules possess nontotally symmetric vibrations that break the maximum hardness (MHP) and minimum polarizability principles (MPP). We have also derived a set of simple rules to determine a priori without
T Nomiyama
Archives of toxicology, 70(1), 43-50 (1995-01-01)
Changes in bronchoalveolar lavage fluid (BALF) and blood were examined to assess the toxic effects of diborane (B2H6, CAS: 19287-45-7) on the lung. Male Wistar rats were exposed to diborane at 20 ppm (intended concentration) for 4 h (phase I
Ibon Alkorta et al.
Physical chemistry chemical physics : PCCP, 13(31), 14026-14032 (2011-06-24)
An ab initio study of an isomer of diborane(4) [B(2)H(4)] has been carried out at MP2/aug-cc-pVTZ to investigate the ground-state properties of this unusual molecule, a derivative of which has been described in the recent literature. The geometric, electronic and
Z D Zhang et al.
Journal of nanoscience and nanotechnology, 1(2), 153-158 (2003-08-14)
Carbon- and boron-oxide-encapsulated iron nanocapsules have been synthesized by arc discharge in methane (CH4) and diborane (B2H6) atmospheres respectively. The characterization and magnetic properties of carbon- and boron-oxide-encapsulated iron nanocapsules [abbreviated as Fe(C) and Fe(B) respectively] were investigated and compared.
M Rafat et al.
Journal of molecular graphics & modelling, 24(2), 111-120 (2005-06-23)
In this article, we describe and apply an algorithm that visualizes atoms and bonds in molecules and van der Waals complexes, based on the topology of the electron density. The theory of quantum chemical topology defines both atoms and bonds
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service