Skip to Content
Merck
All Photos(2)

Key Documents

462667

Sigma-Aldrich

Lead(II) methanesulfonate solution

50 wt. % in H2O

Synonym(s):

Plumbous methanesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€134.00

€134.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€134.00

About This Item

Linear Formula:
(CH3SO3)2Pb
CAS Number:
Molecular Weight:
397.40
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€134.00


Check Cart for Availability

Request a Bulk Order

reaction suitability

core: lead
reagent type: catalyst

Quality Level

concentration

50 wt. % in H2O

refractive index

n20/D 1.423

density

1.7 g/mL at 25 °C

SMILES string

[PbH2++].CS([O-])(=O)=O.CS([O-])(=O)=O

InChI

1S/2CH4O3S.Pb/c2*1-5(2,3)4;/h2*1H3,(H,2,3,4);/q;;+2/p-2

InChI key

LLABTCPIBSAMGS-UHFFFAOYSA-L

Other Notes

Contains methanesulfonic acid

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1A - Skin Corr. 1 - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service