Skip to Content
Merck
All Photos(1)

Documents

462594

Sigma-Aldrich

Sodium dihydrodimethylaluminate solution

1.0 M in THF: toluene (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2AlH2]Na
CAS Number:
Molecular Weight:
82.06
MDL number:
UNSPSC Code:
12352000

reaction suitability

reagent type: reductant

concentration

1.0 M in THF: toluene (1:1)

bp

65-67 °C

density

0.875 g/mL at 25 °C

SMILES string

[Na+].C[AlH2-]C

Packaging

Packaged under nitrogen in Sure/Seal bottles

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service