Skip to Content
Merck
All Photos(1)

Key Documents

462349

Sigma-Aldrich

Poly(1-decene)

viscosity 50 cSt (40 °C)(lit.)

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€87.90

€87.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€87.90

About This Item

Linear Formula:
[CH2CH[(CH2)7CH3]]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€87.90


Check Cart for Availability

Request a Bulk Order

refractive index

n20/D 1.462

viscosity

50 cSt(40 °C)(lit.)

bp

>316 °C (lit.)

density

0.833 g/mL at 25 °C

SMILES string

C(CCCC)CCCC=C

InChI

1S/C10H20/c1-3-5-7-9-10-8-6-4-2/h3H,1,4-10H2,2H3

InChI key

AFFLGGQVNFXPEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(1-decene) belongs to the class of polyolefins. It is a thermoplastic.[1]

Application

Poly(1-decene) may be used to estimate the pour point (PP) properties of palm oil methyl esters.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymers: a property database. (2008)
The effect of polymers and surfactants on the pour point of palm oil methyl esters.
Sern C H, et al.
European Journal of Lipid Science and Technology, 109(4), 440-444 (2007)

Questions

  1. What is HS CODE

    1 answer
    1. The HS or Foreign Trade Commodity Code is listed on the webpage as 3902.90.0050.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service