Skip to Content
Merck
All Photos(1)

Key Documents

461326

Sigma-Aldrich

Melatonin

≥99.5%

Synonym(s):

N-Acetyl-5-methoxytryptamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N2O2
CAS Number:
Molecular Weight:
232.28
Beilstein:
205542
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99.5%

form

powder

mp

116.5-118 °C (lit.)

SMILES string

COc1ccc2[nH]cc(CCNC(C)=O)c2c1

InChI

1S/C13H16N2O2/c1-9(16)14-6-5-10-8-15-13-4-3-11(17-2)7-12(10)13/h3-4,7-8,15H,5-6H2,1-2H3,(H,14,16)

InChI key

DRLFMBDRBRZALE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ismail Gögenur et al.
Journal of pineal research, 57(1), 10-15 (2014-04-09)
The aim was to examine the effect of perioperative melatonin treatment on clinical cardiac morbidity and markers of myocardial ischemia in patients undergoing elective surgery for abdominal aortic aneurism. Reperfusion injury results in increased cardiac morbidity in patients undergoing surgery
Reuben K Wong et al.
Digestive diseases and sciences, 60(1), 186-194 (2014-08-06)
Probiotics have treatment efficacy in irritable bowel syndrome (IBS), but the exact mechanism remains obscure. One hypothesis is the mediation of melatonin levels, leading to changes in IBS symptoms. The purpose of this study was to evaluate the effects of
Juan R Calvo et al.
Journal of pineal research, 55(2), 103-120 (2013-07-31)
Melatonin is the major secretory product synthesized and secreted by the pineal gland and shows both a wide distribution within phylogenetically distant organisms from bacteria to humans and a great functional versatility. In recent years, a considerable amount of experimental
Paul P Drury et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 34(1), 126-135 (2013-10-10)
Melatonin is a naturally occurring indolamine with mild antioxidant properties that is neuroprotective in perinatal animals. There is limited information on its effects on preterm brain injury. In this study, 23 chronically instrumented fetal sheep received 25 minutes of complete umbilical
Mariano Bizzarri et al.
Expert opinion on therapeutic targets, 17(12), 1483-1496 (2013-09-17)
Compelling evidence has highlighted the complex pleiotropic functions elicited by the melatonin in cancer cells. Melatonin behaves as a 'smart killer', i.e., modulating anti-apoptotic processes in normal cells, and triggering pro-apoptotic signals in cancer cells. Melatonin induces programmed cell death

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service