Skip to Content
Merck
All Photos(2)

Key Documents

460176

Sigma-Aldrich

Poly(ethylene glycol) monooleate

average Mn ~860

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€59.10

€59.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€59.10

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€59.10


Check Cart for Availability

Request a Bulk Order

form

solid

mol wt

average Mn ~860

refractive index

n20/D 1.468

bp

>260 °C (lit.)

acid number

4 KOH/g

hydroxyl value

62 mg KOH/g

iodine value

28

solubility

propylene glycol: insoluble
toluene, ethanol, acetone and water: soluble

density

1.034 g/mL at 25 °C

SMILES string

OCCO.[H]\C(CCCCCCCC)=C(/[H])CCCCCCCC(O)=O

InChI

1S/C20H38O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(22)23-19-18-21/h9-10,21H,2-8,11-19H2,1H3/b10-9-

InChI key

MUHFRORXWCGZGE-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Plasticizer, surfactant.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alexander Mietke et al.
Biophysical journal, 109(10), 2023-2036 (2015-11-21)
Cell stiffness is a sensitive indicator of physiological and pathological changes in cells, with many potential applications in biology and medicine. A new method, real-time deformability cytometry, probes cell stiffness at high throughput by exposing cells to a shear flow

Questions

1–2 of 2 Questions  
  1. Can this product be used as a lubricant for centrifuge buckets/load bearing bolts in a GMP setting?

    1 answer
    1. This product is of low M-Clarity Quality Level and may not be suitable for use in a Good Manufacturing Practice setting, as most Good Manufacturing Practice regulated products are of high M-Clarity Quality Levels 500 - 600.

      Helpful?

  2. What does it mean by average Mn ~860 in the description?

    1 answer
    1. This product demonstrates an average Mn of approximately 860 daltons. The ‘average Mn' is the Number Average Molecular Weight of all the polymer chains present in a given sample. The number average molecular weight (Mn) is calculated using the following equation:
      Mn = ∑iNiMi / ∑iNi
      where Mi is the molecular weight of a chain
      where Ni is the number of chains of that molecular weight
      where i is the number of polymer molecules
      Please see the link below to review additional information that may be of interest:
      https://www.sciencedirect.com/topics/engineering/number-average-molecular-weight

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service