Skip to Content
Merck
All Photos(1)

Key Documents

460095

Sigma-Aldrich

Bis(2-butoxyethyl) phthalate

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-[CO2CH2CH2O(CH2)3CH3]2
CAS Number:
Molecular Weight:
366.45
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

technical grade

impurities

10% polyalkyl substitution groups

refractive index

n20/D 1.486 (lit.)

bp

220 °C (lit.)

solubility

hexane, toluene, ethanol and acetone: soluble
water and propylene glycol: insoluble

density

1.06 g/mL at 25 °C (lit.)

SMILES string

CCCCOCCOC(=O)c1ccccc1C(=O)OCCOCCCC

Application

Plasticizer

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santino Orecchio et al.
Journal of toxicology and environmental health. Part A, 78(15), 1008-1018 (2015-08-12)
A simple and fast method is proposed to analyze commercial personal perfumes. Our method includes measurement of phthalates, known to be major sources of endocrine disruptor chemicals (EDC), which originate from the less volatile fraction of perfumes. The quantification of
Wei Jia et al.
Journal of chromatography. A, 1362, 110-118 (2014-08-27)
A new analytical method was developed and validated for simultaneous analysis of 27 phthalates in milk and milk products. Response surface methodology was employed to optimize a quick, easy, cheap, effective, rugged, and safe (QuEChERS) sample preparation method. Ultrahigh-performance liquid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service