Skip to Content
Merck
All Photos(1)

Documents

459267

Sigma-Aldrich

(1S,2S)-(+)-trans-1,2-Cyclopentanediol

98%

Synonym(s):

(1S)-trans-1,2-Cyclopentanediol, (1S,2S)-trans-1,2-Cyclopentanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H8(OH)2
CAS Number:
Molecular Weight:
102.13
Beilstein:
2496900
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D +21.0°, c = 1 in chloroform

optical purity

ee: 97% (GLC)

bp

136 °C/21.5 mmHg (lit.)

mp

47-51 °C (lit.)

SMILES string

O[C@H]1CCC[C@@H]1O

Application

C2 symmetric chiral diol with versatile applications as a chiral auxiliary, building block, and chiral ligand.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stolle, A. et al.
Tetrahedron Letters, 35, 3521-3521 (1994)
Marshall, J.A. Xie, S.
The Journal of Organic Chemistry, 60, 7230-7230 (1995)
Burk, M.J. et al.
Journal of the American Chemical Society, 115, 10125-10125 (1993)
Burk, M.J. et al.
Tetrahedron Asymmetry, 2, 569-569 (1991)
Pichon, M. Figadere, B.
Tetrahedron Asymmetry, 7, 927-927 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service