Skip to Content
Merck
All Photos(1)

Key Documents

459070

Sigma-Aldrich

Potassium hexachloroosmate(IV)

≥99.99% trace metals basis

Synonym(s):

Osmium potassium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2OsCl6
CAS Number:
Molecular Weight:
481.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

≥99.99% trace metals basis

form

solid

mp

600 °C (dec.) (lit.)

density

3.42 g/mL at 25 °C (lit.)

SMILES string

[K+].[K+].Cl[Os--](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.2K.Os/h6*1H;;;/q;;;;;;2*+1;+4/p-6

InChI key

VGKQJCSDERXWRV-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service