Skip to Content
Merck
All Photos(1)

Key Documents

457604

Sigma-Aldrich

β-Propiolactone

97%

Synonym(s):

3-Hydroxypropionic acid lactone, Hydracrylic acid β-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4O2
CAS Number:
Molecular Weight:
72.06
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

97%

refractive index

n20/D 1.412 (lit.)

bp

162 °C (lit.)

mp

−33 °C (lit.)

density

1.146 g/mL at 25 °C (lit.)

SMILES string

O=C1CCO1

InChI

1S/C3H4O2/c4-3-1-2-5-3/h1-2H2

InChI key

VEZXCJBBBCKRPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

During storage insolubles may form due to self-polymerization

Warning

OSHA-regulated carcinogen - 29 CFR Part 1910.1013

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service