Skip to Content
Merck
All Photos(1)

Key Documents

457418

Sigma-Aldrich

5-Bromo-2,4-dimethoxypyrimidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7BrN2O2
CAS Number:
Molecular Weight:
219.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

62-65 °C (lit.)

functional group

bromo

SMILES string

COc1ncc(Br)c(OC)n1

InChI

1S/C6H7BrN2O2/c1-10-5-4(7)3-8-6(9-5)11-2/h3H,1-2H3

InChI key

QEZIMQMMEGPYTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1016. Reactions of pyrimidin-5-yl-lithium compounds.
Caton MPL, et al.
Journal of the Chemical Society, 5467-5473 (1965)
The Hilbert-Johnson Reaction of 2, 4-Dialkoxypyrimidines with Halogenoses.
Pliml J and Prystas M.
Advances in Heterocyclic Chemistry, 8, 115-142 (1967)
Palladium-catalysed carbomethoxyvinylation and thienylation of 5-iodo (bromo)-2, 4-dimethoxypyrimidine in water.
Basnak I, et al.
Tetrahedron Letters, 38(27), 4869-4872 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service