Skip to Content
Merck
All Photos(3)

Key Documents

454648

Sigma-Aldrich

4-tert-Butylphthalic anhydride

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€229.00

€229.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€229.00

About This Item

Empirical Formula (Hill Notation):
C12H12O3
CAS Number:
Molecular Weight:
204.22
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€229.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

form

powder, crystals or chunks

mp

70-75 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC(C)(C)c1ccc2C(=O)OC(=O)c2c1

InChI

1S/C12H12O3/c1-12(2,3)7-4-5-8-9(6-7)11(14)15-10(8)13/h4-6H,1-3H3

InChI key

YLJYVKLZVHWUCT-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
411930374776330590
form

powder, crystals or chunks

form

powder, crystals or chunks

form

powder, crystals or chunks

form

powder, crystals or chunks

assay

95%

assay

98%

assay

97%

assay

99%

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

mp

70-75 °C (lit.)

mp

98-100 °C (lit.)

mp

86-90 °C (lit.)

mp

142-144 °C (lit.)

Application

4-tert-Butylphthalic anhydride has been used to synthesize novel yellow quinophthalone dyes for LCD color filters.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service