454419
Ethyldiphenylphosphine oxide
97%
Synonym(s):
Diphenylphosphinoethane monoxide
About This Item
Recommended Products
Assay
97%
form
solid
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
mp
120-125 °C (lit.)
functional group
phosphine oxide
SMILES string
CCP(=O)(c1ccccc1)c2ccccc2
InChI
1S/C14H15OP/c1-2-16(15,13-9-5-3-6-10-13)14-11-7-4-8-12-14/h3-12H,2H2,1H3
InChI key
AKTGKEBIBGSCLD-UHFFFAOYSA-N
Application
- Regioselective lactonization of unsymmetrical 1,4-diols
- Synthesis of a nanostructured dioxomolybdenum(VI) catalyst for liquid-phase epoxidation of olefins
- Reduction of tertiary phosphine oxides with DIBAL-H
- Diphenylphosphineyl-mediated synthesis of ketones
- Nucleophilic aromatic substitution of hydrogen by phosphorous stabilized carbanions
- Stereospecific reduction to form phosphines
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service