All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H8N2O
CAS Number:
Molecular Weight:
136.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
95%
mp
163-169 °C (lit.)
functional group
nitrile
SMILES string
Cc1oc(N)c(C#N)c1C
InChI
1S/C7H8N2O/c1-4-5(2)10-7(9)6(4)3-8/h9H2,1-2H3
InChI key
ACHDPRAJHGRGDC-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
2-Amino-3-cyano-4, 5-dimethylfuran as precursors to butenolides and arylacetic acids.
Kooshkabadi H, et al.
Heterocycles, 29(7), 1221-1224 (1989)
N A Vaidya et al.
Journal of medicinal chemistry, 26(10), 1422-1425 (1983-10-01)
Twenty-three substituted 3,4-dihydro-4-oxoquinazolines or 3,4-dihydro-4-oxoazaquinazolines have been synthesized utilizing 2-amino-3-cyano-4,5-dimethylfuran and methyl acrylate as precursors for synthesis of the required substituted anthranilates. Six additional azaquinazolones were synthesized from 2-aminonicotinic or 3-aminopicolinic acid for comparison studies. All compounds were evaluated in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service