Skip to Content
Merck
All Photos(1)

Key Documents

451835

Sigma-Aldrich

Bisphenol A-d16

98 atom % D

Synonym(s):

2,2-·Bis(4-·hydroxyphenyl)·propane-D16, 4,4·-Isopropylidenediphenol-D16, Deuterated bisphenol A, 2,2-Bis(4-hydroxyphenyl)propane-d16

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€146.00
250 MG
€418.00
1 G
€706.00

€146.00


Check Cart for Availability

Request more information

Select a Size

Change View
50 MG
€146.00
250 MG
€418.00
1 G
€706.00

About This Item

Linear Formula:
(CD3)2C(C6D4OD)2
CAS Number:
Molecular Weight:
244.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12

€146.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

bp

220 °C/4 mmHg (lit.)

mp

157-159 °C (lit.)

mass shift

M+16

storage temp.

room temp

SMILES string

[2H]Oc1c([2H])c([2H])c(c([2H])c1[2H])C(c2c([2H])c([2H])c(O[2H])c([2H])c2[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17H,1-2H3/i1D3,2D3,3D,4D,5D,6D,7D,8D,9D,10D/hD2

InChI key

IISBACLAFKSPIT-MAJJRYNQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

May be used as an analytical standard

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Analysis Note

This product may be used as an analytical standard.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruna De Felice et al.
BMC medical genomics, 8, 56-56 (2015-09-09)
Bisphenol A (BPA) is an environmental compounds is known to possess endocrine disruption potentials. Bisphenol A has epigenetic effects as deregulated expression of microRNAs; such epigenetic marks can induce up/down alterations in gene expression that may persist throughout a lifetime.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service