Skip to Content
Merck
All Photos(1)

Key Documents

450251

Sigma-Aldrich

Hydrazine dihydrochloride

≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2 · 2HCl
CAS Number:
Molecular Weight:
104.97
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.99% trace metals basis

form

solid

mp

200 °C (dec.) (lit.)

density

1.42 g/mL at 25 °C (lit.)

SMILES string

Cl[H].Cl[H].NN

InChI

1S/2ClH.H4N2/c;;1-2/h2*1H;1-2H2

InChI key

LIAWOTKNAVAKCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic-spectrophotometric determination of molybdenum in plant and steel with nile red-hydrazine dihydrochloride reaction.
Ensafi AA.
Analytical Letters, 25(!2), 2339-2354 (1992)
Spectrophotometric method for determination of hydrazine.
Watt GW and Chrisp JD.
Analytical Chemistry, 24(12), 2006-2008 (1952)
Accelerated synthesis of 3, 5-disubstituted 4-amino-1, 2, 4-triazoles under microwave irradiation.
Bentiss F, et al.
Tetrahedron Letters, 41(10), 1539-1541 (2000)
Rapid synthesis of 2, 5-disubstituted 1, 3, 4-oxadiazoles under microwave irradiation.
Bentiss F, et al.
Synthetic Communications, 31(6), 935-938 (2001)
A silver colloid produced by reduction with hydrazine as support for highly sensitive surface-enhanced Raman spectroscopy
Nickel U, et al.
Langmuir, 16(23), 9087-9091 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service