Skip to Content
Merck
All Photos(2)

Key Documents

450219

Sigma-Aldrich

Silver hexafluorophosphate

99.99% trace metals basis

Synonym(s):

Silver hexafluorophosphate (AgPF6 ), Silver(1+) hexafluorophosphate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€75.90
5 G
€267.00

€75.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€75.90
5 G
€267.00

About This Item

Linear Formula:
AgPF6
CAS Number:
Molecular Weight:
252.83
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

€75.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: silver

impurities

≤150.0 ppm Trace Metal Analysis

mp

102 °C (dec.) (lit.)

SMILES string

[Ag+].F[P-](F)(F)(F)(F)F

InChI

1S/Ag.F6P/c;1-7(2,3,4,5)6/q+1;-1

InChI key

SCQBROMTFBBDHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in the development of new catalyst systems for stereoselective addition of alcohols to alkynes.[1] Acidic catalyst for the synthesis of unsymmetrical sulfides[2] and vinyl fluorides.[3] Used to prepare silver 3,3′-dicyanodiphenylacetylene coordination networks for study of the effect of ligands on network structure toward the goal of engineering novel materials.[4]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic Communications, 22, 1461-1461 (1992)
Chemistry Letters (Jpn), 727-727 (1996)
Chemistry Letters (Jpn), 1-1 (1993)
Keith A. Hirsch et al.
Inorganic chemistry, 36(14), 2960-2968 (1997-07-02)
Coordination networks of 3,3'-dicyanodiphenylacetylene (3,3'-DCPA, 1) with silver(I) salts characterized by single-crystal X-ray analysis are described. Network topology is found to depend on both the counterion and solvent employed during crystallization. The conformation adopted by the ligand varies between planar

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service