Skip to Content
Merck
All Photos(1)

Key Documents

450189

Sigma-Aldrich

Lithium acetate dihydrate

99.999% trace metals basis

Synonym(s):

Acetic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOLi · 2H2O
CAS Number:
Molecular Weight:
102.02
Beilstein:
3564320
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.999% trace metals basis

form

solid

reaction suitability

core: lithium

mp

53-56 °C (lit.)

SMILES string

[Li+].[H]O[H].[H]O[H].CC([O-])=O

InChI

1S/C2H4O2.Li.2H2O/c1-2(3)4;;;/h1H3,(H,3,4);;2*1H2/q;+1;;/p-1

InChI key

IAQLJCYTGRMXMA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zico Alaia Akbar et al.
Optics express, 23(19), A1280-A1287 (2015-09-26)
A unique, hierarchically structured, aggregated TiO(2) nanowire (A-TiO(2)-nw) is prepared by solvothermal synthesis and used as a dual-functioning photoelectrode in dye-sensitized solar cells (DSSCs). The A-TiO(2)-nw shows improved light scattering compared to conventional TiO(2) nanoparticles (TiO(2)-np) and dramatically enhanced dye
Tomasz Krawczyk
Rapid communications in mass spectrometry : RCM, 29(23), 2257-2262 (2015-11-03)
Hexamethylene triperoxide diamine (HMTD) is one of the peroxide-based explosives that are difficult to detect using standard analytical methodologies. It was analyzed by electrospray ionization mass spectrometry (ESI-MS) on a UPLC-TOF instrument. Alkali metal salts were used to promote the
Wenyi Hua et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 969, 117-122 (2014-08-30)
A highly sensitive, selective, and rugged quantification method was developed and validated for decitabine (5-aza-2'-deoxycytidine) in human plasma treated with 100μg/mL of tetrahydrouridine (THU). Chromatographic separation was accomplished using hydrophilic interaction liquid chromatography (HILIC) and detection used electrospray ionization (ESI)
Elaine L Ferguson et al.
Molecular pharmaceutics, 11(12), 4437-4447 (2014-11-02)
Polymer therapeutics offer potential benefits in the treatment of multidrug resistant (MDR) infections; affording targeted delivery of biologically active agents to the site of inflammation, potential decreases in systemic toxicity, and the retention of antimicrobial activity at the target site.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service