Skip to Content
Merck
All Photos(1)

Key Documents

450154

Sigma-Aldrich

Cesium acetate

≥99.99% trace metals basis

Synonym(s):

Cesium Ethanoate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€195.00

€195.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€195.00

About This Item

Linear Formula:
CH3COOCs
CAS Number:
Molecular Weight:
191.95
Beilstein:
3595637
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€195.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.99% trace metals basis

form

solid

mp

194 °C (lit.)

SMILES string

[Cs+].CC([O-])=O

InChI

1S/C2H4O2.Cs/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

ZOAIGCHJWKDIPJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Facile synthesis of 1′,2′-cis-beta-pyranosyladenine nucleosides.: This study presents a simplified method for synthesizing beta-pyranosyladenine nucleosides, potentially useful in nucleic acid research and drug development. Cesium acetate is highlighted as a key reagent in promoting glycosylation reactions, demonstrating its utility in organic synthesis (Ando et al., 2007).


  • Raman spectroscopy and dioxygen adsorption on Cs-loaded zeolite catalysts for butene isomerization.: Investigates the role of cesium acetate in enhancing the catalytic properties of zeolites for butene isomerization. The study uses Raman spectroscopy to elucidate the interaction between cesium ions and the zeolite framework, offering insights into the design of more efficient catalysts (Li and Davis, 2005).


Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Xin et al.
Small (Weinheim an der Bergstrasse, Germany), 16(43), e2004513-e2004513 (2020-10-03)
High-energy radiation detectors such as X-ray detectors with low light photoresponse characteristics are used for several applications including, space, medical, and military devices. Here, an indirect bandgap inorganic perovskite-based X-ray detector is reported. The indirect bandgap nature of perovskite materials
Zhaoyan Yang et al.
ACS applied materials & interfaces, 11(51), 47671-47679 (2019-10-22)
Lead halide perovskite nanocrystals (NCs) are promising optical materials in many fields. However, their poor moisture stability, significant toxicity, and difficulty to be further functionalized greatly hinder their applications in bioimaging. Here, a universal strategy is demonstrated by simply encapsulating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service