Skip to Content
Merck
All Photos(1)

Key Documents

448680

Sigma-Aldrich

α-Ethylbenzylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(C2H5)NH2
CAS Number:
Molecular Weight:
135.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

refractive index

n20/D 1.519 (lit.)

bp

204 °C (lit.)

density

0.938 g/mL at 25 °C (lit.)

functional group

amine
phenyl

SMILES string

CCC(N)c1ccccc1

InChI

1S/C9H13N/c1-2-9(10)8-6-4-3-5-7-8/h3-7,9H,2,10H2,1H3

InChI key

AQFLVLHRZFLDDV-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
126993M31201631086
Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

-

density

0.938 g/mL at 25 °C (lit.)

density

0.909 g/mL at 25 °C (lit.)

density

0.952 g/mL at 25 °C (lit.)

density

0.951 g/mL at 25 °C (lit.)

refractive index

n20/D 1.519 (lit.)

refractive index

n20/D 1.511 (lit.)

refractive index

n20/D 1.534 (lit.)

refractive index

n20/D 1.5410 (lit.)

bp

204 °C (lit.)

bp

191-194 °C (lit.)

bp

195 °C (lit.)

bp

103-106 °C/0.5 mmHg (lit.)

functional group

amine

functional group

amine, phenyl

functional group

-

functional group

amine

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalysis with supported palladium metal, selectivity in the hydrogenation of C? C, C? O and C? N bonds, from chemo-to enantioselectivity.
Tungler A and Fogassy G.
J. Mol. Catal. A: Chem., 173(1), 231-247 (2001)
Optical resolution of a-ethylbenzylamine and its application as a resolving agent.
Nohira H, et al.
Bulletin of the Chemical Society of Japan, 61(4), 1395-1396 (1988)
Yujie Chen et al.
Applied microbiology and biotechnology, 100(7), 3101-3111 (2015-11-19)
ω-Transaminases (ω-TAs) are one of the most popular candidate enzymes in the biosynthesis of chiral amines. Determination of yet unidentified ω-TAs is important to broaden their potential for synthetic application. Taurine-pyruvate TA (TPTA, EC 2.6.1.77) is an ω-TA belonging to
Kuei Hui Chan et al.
Biomedical chromatography : BMC, 19(5), 337-342 (2005-06-03)
A rapid and reliable high-performance liquid chromatographic method was developed and validated for the simultaneous determination of norephedrine (NE), norpseudoephedrine (NPE), ephedrine (E), pseudoephedrine (PE), methylephedrine (ME) and methylpseudoephedrine (MPE) in both a Mahwang traditional Chinese medicine (TCM) preparation and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service