Skip to Content
Merck
All Photos(1)

Documents

448060

Sigma-Aldrich

Arsenic(III) sulfide

≥99.99%

Synonym(s):

Arsenic sesquisulfide, Arsenic sulfide, Arsenic trisulfide, Arsenic trisulfide (As2 S3 ), Arsenious sulfide, Diarsenic trisulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
As2S3
CAS Number:
Molecular Weight:
246.04
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥99.99%

mp

300 °C (lit.)

density

3.43 g/mL at 25 °C (lit.)

SMILES string

S=[As]S[As]=S

InChI

1S/As2S3/c3-1-5-2-4

InChI key

KNFJTZYMLKHMCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Arsenic trisulfide is a strong reducing agent. It can be dissolved in strong acids and bases.3 It has antitumor and anticancer properties.

Application

Arsenic sulfide may be used in the semiconductor industry

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M El-Amraoui et al.
Optics express, 18(25), 26655-26665 (2010-12-18)
The aim of this paper is to present an overview of the recent achievements of our group in the fabrication and optical characterizations of As(2)S(3) microstructured optical fibers (MOFs). Firstly, we study the synthesis of high purity arsenic sulfide glasses.
S Focardi et al.
Letters in applied microbiology, 51(5), 578-585 (2010-09-24)
To isolate and characterize an anaerobic bacterial strain from the deeper polluted lagoon sediment able to use as electron acceptors [As(V)] and sulfate (SO4(2-)), using lactate as an electron donor. Methods for isolation from polluted lagoon sediments included anaerobic enrichment
Dong-Hun Kim et al.
Journal of bacteriology, 193(18), 5039-5040 (2011-08-27)
The dissimilatory metal reducing bacterium Shewanella sp. strain HN-41 was first reported to produce novel photoactive As-S nanotubes via reduction of As(V) and S(2)O(3)(2-) under anaerobic conditions. Here we report the draft genome sequence and annotation of strain HN-41.
R J Weiblen et al.
Optics express, 18(25), 26666-26674 (2010-12-18)
We computationally investigate supercontinuum generation in an As(2)S(3) solid core photonic crystal fiber (PCF) with a hexagonal cladding of air holes. We study the effect of varying the system (fiber and input pulse) parameters on the output bandwidth. We find
M D Pelusi et al.
Optics express, 18(25), 26686-26694 (2010-12-18)
We report the first demonstration of optical phase conjugation (OPC) transmission of phase encoded and wavelength-division multiplexed (WDM) signals by the Kerr effect in a planar structured waveguide. The phase conjugated electric field of the signal is produced by four

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service