Skip to Content
Merck
All Photos(1)

Key Documents

447277

Sigma-Aldrich

Allyl(dichloro)methylsilane

95%

Synonym(s):

Allyl-methyl-dichlorosilane

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€445.00

€445.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€445.00

About This Item

Linear Formula:
H2C=CHCH2Si(Cl)2CH3
CAS Number:
Molecular Weight:
155.10
Beilstein:
1740877
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€445.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

refractive index

n20/D 1.440 (lit.)

bp

119.5 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)

SMILES string

C[Si](Cl)(Cl)CC=C

InChI

1S/C4H8Cl2Si/c1-3-4-7(2,5)6/h3H,1,4H2,2H3

InChI key

YCEQUKAYVABWTE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self-Folded Silyl Cavitands with In-and Outwardly Directed Allyl Groups.
Ohashi K, et al.
European Journal of Organic Chemistry, 8, 1597-1601 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service