Skip to Content
Merck
All Photos(1)

Key Documents

447110

Sigma-Aldrich

Tetrabutylammonium phosphate monobasic solution

0.4 M in acetonitrile

Synonym(s):

Tetrabutylammonium dihydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N[OP(OH)2O]
CAS Number:
Molecular Weight:
339.45
Beilstein:
5196532
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

concentration

0.4 M in acetonitrile

refractive index

n20/D 1.369

bp

81-82 °C

density

0.828 g/mL at 25 °C

SMILES string

OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H3O4P/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H3,1,2,3,4)/q+1;/p-1

InChI key

ARRNBPCNZJXHRJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service