All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H12
CAS Number:
Molecular Weight:
192.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
mp
157-161 °C (lit.)
functional group
phenyl
SMILES string
C1c2ccccc2C=C1c3ccccc3
InChI
1S/C15H12/c1-2-6-12(7-3-1)15-10-13-8-4-5-9-14(13)11-15/h1-10H,11H2
InChI key
BSBXLZYWGGAVHD-UHFFFAOYSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A Convenient Synthesis of 2-Phenylindene.
Jayamani M and Pillai CN.
Synthetic Communications, 15(6), 535-542 (1985)
Observation of intramolecular vibrational redistribution and vibrational cooling in S 1 trans-stilbene and 2-phenylindene in solution.
Qian J, et al.
Chemical Physics Letters, 233(1), 9-15 (1995)
Chemical behavior of o-bis (phenylethynyl) benzene toward some electrophilic and nucleophilic reagents.
Whitlock Jr HW, et al.
The Journal of Organic Chemistry, 34(4), 879-886 (1969)
Christian Müller et al.
The journal of physical chemistry. A, 110(15), 5017-5031 (2006-04-14)
We investigated the spectroscopy of the first excited singlet electronic state S1 of 2-phenylindene using both fluorescence excitation spectroscopy and resonantly enhanced multiphoton ionization spectroscopy. Moreover, we investigated the dynamics of the S1 state by determining state-selective fluorescence lifetimes up
Ozonolyses of 1, 2-diphenyl-and 2-phenylindene.
Sugimoto T, et al.
The Journal of Organic Chemistry, 58(1), 135-141 (1993)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service