Skip to Content
Merck
All Photos(1)

Key Documents

444529

Sigma-Aldrich

Zinc 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-hexadecafluoro-29H,31H-phthalocyanine

Dye content 90 %

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€201.00

€201.00


Estimated to ship on29 March 2025


Request a Bulk Order

Select a Size

Change View
1 G
€201.00

About This Item

Empirical Formula (Hill Notation):
C32F16N8Zn
CAS Number:
Molecular Weight:
865.76
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€201.00


Estimated to ship on29 March 2025


Request a Bulk Order

form

powder

Quality Level

composition

Dye content, 90%

mp

>300 °C (lit.)

λmax

686 nm

SMILES string

Fc1c(F)c(F)c2c3nc(nc4n5[Zn]n6c(n3)c7c(F)c(F)c(F)c(F)c7c6nc8nc(nc5c9c(F)c(F)c(F)c(F)c49)c%10c(F)c(F)c(F)c(F)c8%10)c2c1F

InChI

1S/C32F16N8.Zn/c33-9-1-2(10(34)18(42)17(9)41)26-49-25(1)53-27-3-4(12(36)20(44)19(43)11(3)35)29(50-27)55-31-7-8(16(40)24(48)23(47)15(7)39)32(52-31)56-30-6-5(28(51-30)54-26)13(37)21(45)22(46)14(6)38;/q-2;+2

InChI key

BEAYCAVYGQXGEJ-UHFFFAOYSA-N

Application

Used in a study of photodynamic tumor therapy.[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R W Boyle et al.
British journal of cancer, 73(1), 49-53 (1996-01-01)
Hexadecafluorinated zinc phthalocyanine (ZnPcF16), an analogue of zinc phthalocyanine (ZnPc) in which all hydrogen atoms have been substituted by fluorine, was prepared as a single isomeric product via the condensation of tetrafluorophthalonitrile with zinc acetate. Fluorination renders the ZnPc soluble
E Allémann et al.
International journal of cancer, 66(6), 821-824 (1996-06-11)
Hexadecafluoro zinc phthalocyanine (ZnPcF16), a second-generation sensitizer for the photodynamic therapy (PDT) of cancer, was formulated in polyethylene-glycol-coated poly(lactic acid) nanoparticles (PEG-coated PLA-NP) and tested in EMT-6 tumour-bearing mice for its photodynamic activity. The tumour response was compared to that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service