Skip to Content
Merck
All Photos(1)

Key Documents

444332

Sigma-Aldrich

2-(tert-Butylamino)ethyl methacrylate

97%, contains ~1000 ppm monomethyl ether hydroquinone (MEHQ) as inhibitor

Synonym(s):

2-(N -tert -Butylamino)ethyl methacrylate, 2-(tert -Butylamino)ethyl methacrylate, N -tert -Butylaminoethyl methacrylate, t-Butylaminoethyl methacrylate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€58.20
500 ML
€133.00

€58.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€58.20
500 ML
€133.00

About This Item

Linear Formula:
H2C=C(CH3)CO2CH2CH2NHC(CH3)3
CAS Number:
Molecular Weight:
185.26
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€58.20


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

contains

~1000 ppm monomethyl ether hydroquinone (MEHQ) as inhibitor

refractive index

n20/D 1.442 (lit.)

bp

82 °C/10 mmHg (lit.)

density

0.914 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCCNC(C)(C)C

InChI

1S/C10H19NO2/c1-8(2)9(12)13-7-6-11-10(3,4)5/h11H,1,6-7H2,2-5H3

InChI key

BEWCNXNIQCLWHP-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lingyan Ren et al.
Journal of nanoscience and nanotechnology, 19(9), 5774-5782 (2019-04-10)
In this work, we investigated the influence of different concentrations of silver 2-ethylhexanoate on the synthesis and characteristics of silver nanoparticles (AgNPs) in resin matrix, as well as the antibacterial performance, mechanical properties and biocompatibility of the silver containing resin
Lei Cheng et al.
International journal of oral science, 8(3), 172-181 (2016-06-10)
Dental composites are commonly used restorative materials; however, secondary caries due to biofilm acids remains a major problem. The objectives of this study were (1) to develop a composite containing quaternary ammonium dimethacrylate (QADM), nanoparticles of silver (NAg), and nanoparticles
Sho Hirai et al.
Polymers, 12(11) (2020-11-22)
The consumption of polypropylene (PP) has significantly increased over that of other materials because of its light weight, easy molding, and high mechanical strength. However, the applications of PP are limited, owing to the lack of surface properties, especially with
Chien-Lin Huang et al.
Polymers, 12(7) (2020-07-02)
A novel cationic polymer, poly[2-(tert-butylaminoethyl) methacrylate] (PTA), effectively kills various strains of bacteria with low toxicity to tissue cells. Graphene-based materials demonstrate exceptional electron transport capability, antibacterial activity, favorable nontoxicity, and versatile applicability. PTA can be grafted onto the graphene

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service