Skip to Content
Merck
All Photos(2)

Key Documents

444162

Sigma-Aldrich

Barium iodate monohydrate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€194.00

€194.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€194.00

About This Item

Linear Formula:
Ba(IO3)2 · H2O
CAS Number:
Molecular Weight:
505.15
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
grade:
for analytical purposes
form:
solid

€194.00


Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Assay

98%

form

solid

SMILES string

O.[Ba++].[O-]I(=O)=O.[O-]I(=O)=O

InChI

1S/Ba.2HIO3.H2O/c;2*2-1(3)4;/h;2*(H,2,3,4);1H2/q+2;;;/p-2

InChI key

JFEZLZKLXGWZEC-UHFFFAOYSA-L

Related Categories

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service