Skip to Content
Merck
All Photos(1)

Documents

441716

Sigma-Aldrich

Bis[1-(tert-butylperoxy)-1-methylethyl]benzene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4[C(CH3)2OOC(CH3)3]2
CAS Number:
Molecular Weight:
338.48
EC Number:
MDL number:
UNSPSC Code:
12162002

Assay

96%

composition

active oxygen content, 9 wt. %

t1/2

10 hr(110 °C)

mp

44-48 °C (lit.)
≥80 °C (SADT) (lit.)

SMILES string

CC(C)(C)OOC(C)(C)c1cccc(c1)C(C)(C)OOC(C)(C)C

Application

Free radical polymerization initiator.
Initiates degradation of polypropylene, initiates polymerization and cross-links elastomers and olefins. Acts as a synergist in halogen-containing fire retardant plastics.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Org. Perox. D

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service