Skip to Content
Merck
All Photos(1)

Key Documents

441597

Sigma-Aldrich

D-Asparagine

99%, for peptide synthesis

Synonym(s):

(R)-2-Aminosuccinamic acid, (R)-2-Aminosuccinic acid 4-amide, D-Aspartic acid 4-amide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€334.00

€334.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€334.00

About This Item

Linear Formula:
H2NCOCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
132.12
Beilstein:
1723526
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€334.00


Check Cart for Availability

Request a Bulk Order

Product Name

D-Asparagine, 99%

Quality Level

Assay

99%

form

solid

optical activity

[α]20/D −35°, c = 5 in 5 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

mp

280 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

N[C@H](CC(N)=O)C(O)=O

InChI

1S/C4H8N2O3/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H2,6,7)(H,8,9)/t2-/m1/s1

InChI key

DCXYFEDJOCDNAF-UWTATZPHSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M Michalak et al.
Cell death and differentiation, 15(6), 1019-1029 (2008-02-09)
The ability of p53 to induce apoptosis in cells with damaged DNA is thought to contribute greatly to its tumour suppressor function. P53 has been proposed to induce apoptosis via numerous transcriptional targets or even by direct cytoplasmic action. Two

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service