All Photos(2)
About This Item
Linear Formula:
C6H4(COC6H4F)2
CAS Number:
Molecular Weight:
322.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
98%
form
solid
mp
181-183 °C (lit.)
functional group
fluoro
ketone
SMILES string
Fc1ccc(cc1)C(=O)c2cccc(c2)C(=O)c3ccc(F)cc3
InChI
1S/C20H12F2O2/c21-17-8-4-13(5-9-17)19(23)15-2-1-3-16(12-15)20(24)14-6-10-18(22)11-7-14/h1-12H
InChI key
PISLKPDKKIDMQT-UHFFFAOYSA-N
General description
1,3-Bis(4-fluorobenzoyl)benzene is an organic building block. It polymerizes with 4,4′-isopropylidenediphenol (bisphenol-A) in the presence of N-methyl-2-pyrrolidinone (solvent) to afford high molecular weight polymers.[1]
Application
1,3-Bis(4-fluorobenzoyl)benzene was used for the synthesis of sulfonated poly(aryl ether ketone)s (SPAEK) copolymers by aromatic nucleophilic polycondensation.[2] It may be employed as an activated nonsulfonated difluoro-monomer for the synthesis of fluorenyl-containing sulfonated poly(aryl ether ether ketone ketone)s.[3]
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Polyimides containing carbonyl and ether connecting groups.
Hergenrother PM, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 25(4), 1093-1103 (1987)
Fluorenyl-containing sulfonated poly (aryl ether ether ketone ketone) s (SPFEEKK) for fuel cell applications
Li B, et al.
Journal of Membrane Science, 280(1), 54-64 (2006)
Macromolecular Chemistry and Physics, 197, 315-315 (1996)
Synthesis and characterization of poly (aryl ether ketone) copolymers containing (hexafluoroisopropylidene)-diphenol moiety as proton exchange membrane materials.
Xing P, et al.
Polymer, 46(10), 3257-3263 (2005)
Competing processes for poly (aryl ether-ketone) synthesis, 3. Polymerization reactions of 1, 3-bis (4-chlorobenzoyl) benzene and 1, 3-bis (4-fluorobenzoyl) benzene in N-methyl-2-pyrrolidinone.
Bhatnagar A, et al.
Macromolecular Chemistry and Physics, 197(1), 315-328 (1996)
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service