Skip to Content
Merck
All Photos(1)

Key Documents

440299

Sigma-Aldrich

Methyltrichlorosilane

≥96%

Synonym(s):

Trichloro(methyl)silane

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€103.00

€103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€103.00

About This Item

Linear Formula:
CH3SiCl3
CAS Number:
Molecular Weight:
149.48
Beilstein:
1361381
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

€103.00


Check Cart for Availability

Request a Bulk Order

vapor density

5.2 (vs air)

Quality Level

vapor pressure

150 mmHg ( 25 °C)

Assay

≥96%

form

liquid

autoignition temp.

>760 °F

expl. lim.

11.9 %

refractive index

n20/D 1.411 (lit.)

bp

66 °C (lit.)

density

1.273 g/mL at 25 °C (lit.)

SMILES string

C[Si](Cl)(Cl)Cl

InChI

1S/CH3Cl3Si/c1-5(2,3)4/h1H3

InChI key

JLUFWMXJHAVVNN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingbin Ge et al.
The journal of physical chemistry. A, 114(6), 2384-2392 (2010-01-29)
The rate constants for the gas-phase reactions in the silicon carbide chemical vapor deposition of methyltrichlorosilane (Ge, Y. B.; Gordon, M. S.; Battaglia, F.; Fox, R. O. J. Phys. Chem. A 2007, 111, 1462.) were calculated. Transition state theory was
Ana Gisela Cunha et al.
Journal of colloid and interface science, 344(2), 588-595 (2010-02-05)
This work describes a very simple, rapid, and efficient approach to the hydrophobization and lipophobization of cellulose fibers through their reaction with gaseous trichloromethylsilane (TCMS). The characterization of the modified surface involved FTIR-ATR and solid-state (29)Si NMR spectroscopy, scanning electron
Yingbin Ge et al.
The journal of physical chemistry. A, 111(8), 1475-1486 (2007-02-06)
The kinetics for the previously proposed 114-reaction mechanism for the chemical vapor deposition (CVD) process that leads from methyltrichlorosilane (MTS) to silicon carbide (SiC) are examined. Among the 114 reactions, 41 are predicted to proceed with no intervening barrier. For
W H Mullen et al.
Clinica chimica acta; international journal of clinical chemistry, 157(2), 191-198 (1986-06-15)
An enzyme electrode is described based upon the enzyme lactate oxidase (EC 1.1.3.2) coupled to an H2O2 sensor. Use of an organosilane-treated microporous membrane over the enzyme layer, allows responses linear to 18 mmol/l L-lactate with response times of 1-3
Ildikó Szabó et al.
Biopolymers, 88(1), 20-28 (2006-10-26)
Rearrangement of disulfide bonds during the synthesis of alpha-conotoxin GI using PhS(O)Ph/CH(3)SiCl(3) oxidation procedure was observed. We have demonstrated that the protecting scheme (order of acetamidomethyl (Acm) and (t)Bu protecting groups) of the Cys residues as well as the reaction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service