Skip to Content
Merck
All Photos(1)

Key Documents

439525

Sigma-Aldrich

2-Mercaptobenzyl alcohol

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSC6H4CH2OH
CAS Number:
Molecular Weight:
140.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

solid

refractive index

n20/D 1.612 (lit.)

mp

31-32 °C (lit.)

density

1.21 g/mL at 25 °C (lit.)

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

OCc1ccccc1S

InChI

1S/C7H8OS/c8-5-6-3-1-2-4-7(6)9/h1-4,8-9H,5H2

InChI key

FYWFCRHZXORPFH-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Herbert Meier
Molecules (Basel, Switzerland), 17(2), 1548-1570 (2012-02-09)
Benzo-condensed four-ring heterocycles, such as benzoxetes 1 and benzothietes 3 represent multi-purpose starting compounds for the preparation of various higher heterocyclic ring systems. The thermal or photochemical valence isomerizations between the benzenoid forms 1,3 and the higher reactive o-quinoid structures
Synthesis of adjustable poly (vinyl chloride) networks.
Reinecke H, et al.
Macromolecular Rapid Communications, 17(1), 15-23 (1996)
An expedient synthesis of diversified pyrrolizines and indolizines.
Bashiardes G, et al.
Tetrahedron Letters, 45(7), 1567-1570 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service