Skip to Content
Merck
All Photos(1)

Key Documents

438545

Sigma-Aldrich

4-Amino-5-(4-pyridyl)-4H-1,2,4-triazole-3-thiol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€81.50

€81.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€81.50

About This Item

Empirical Formula (Hill Notation):
C7H7N5S
CAS Number:
Molecular Weight:
193.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€81.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

250-254 °C (lit.)

SMILES string

Nn1c(S)nnc1-c2ccncc2

InChI

1S/C7H7N5S/c8-12-6(10-11-7(12)13)5-1-3-9-4-2-5/h1-4H,8H2,(H,11,13)

InChI key

LHSIJUVRXSETDR-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Generalized approach to SERS-active nanomaterials via controlled nanoparticle linking, polymer encapsulation, and small-molecule infusion.
Braun GB, et al.
The Journal of Physical Chemistry C, 113(31), 13622-13629 (2009)
A Silsesquioxane Organically Modified with 4-Amino-5-(4-pyridyl)-4H-1,2,4-triazole-3-thiol: Thermal Behavior and Its Electrochemical Detection of Sulfhydryl Compounds.
Do Carmo DR, et al.
Journal of Nanomaterials, 2014 (2014)
Raghad Haddad et al.
SpringerPlus, 2, 510-510 (2013-10-18)
Series of coordination complexes of Ni(II), Cu(II), Zn(II), Cd(II) and Sn(II) metal with 4-amino-5-(pyridyl)-4H-1,2,4-triazole-3-thiol, as a ligand has been successfully prepared in alcoholic medium. The prepared complexes were characterized quantitatively and qualitatively by using: microelemental analysis, FTIR spectroscopy, UV-visible spectroscopy
A new hybrid polyhedral cubic silsesquioxane chemically modified with 4-amino-5-(4-pyridyl)-4H-1, 2, 4-triazole-3-thiol (APTT) and copper hexacyanoferrate (III) for voltammetric determination of nitrite.
Soares LA, et al.
International Journal of Electrochemical Science, 8, 4654-4669 (2013)
Hamid Reza Akbari Hasanjani et al.
Biosensors & bioelectronics, 128, 1-8 (2019-01-08)
The present work describes an ultrasensitive electrochemical sensor for determination of mercury(II) using deoxyribonucleic acid/poly-L-methionine-gold nanoparticles/pencil graphite electrode (DNA/PMET-AuNPs/PGE). To fabricate this biosensor, L-methionine (L-MET) was electropolymerized on the PGE surface followed by simultaneous electrochemical entrapment of AuNPs. Next, DNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service