Skip to Content
Merck
All Photos(1)

Documents

437026

Sigma-Aldrich

(S)-(+)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione

98%

Synonym(s):

(S)-(+)-8a-Methyl-3,4,8,8a-tetrahydro-1,6(2H,7H)-naphthalenedione, (S)-(+)-9-Methyl-5(10)-octaline-1,6-dione, (S)-Miescher-Wieland ketone, Wieland-Miescher ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14O2
CAS Number:
Molecular Weight:
178.23
Beilstein:
1569979
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D +98°, c = 1.1 in benzene

mp

50-52 °C (lit.)

SMILES string

C[C@]12CCC(=O)C=C1CCCC2=O

InChI

1S/C11H14O2/c1-11-6-5-9(12)7-8(11)3-2-4-10(11)13/h7H,2-6H2,1H3/t11-/m0/s1

InChI key

DNHDRUMZDHWHKG-NSHDSACASA-N

Application

Key intermediate in the total synthesis of the macrocyclic diterpene lactone, (+)-cleomeolide. Chiral building block for enantioselective approaches to the A and C rings of taxol. Used to study selective hydrogenations of the carbon-carbon double bonds in α,β-unsaturated ketones with a new class of palladium complexes as catalysts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arseniyadis, S. et al.
Tetrahedron Letters, 35, 99-99 (1994)
Sommovigo, M. Alper, H.
Tetrahedron Letters, 34, 59-59 (1993)
Paquette, L.A. et al.
Journal of the American Chemical Society, 116, 3367-3367 (1994)
L.A. Paquette et al.
Tetrahedron Letters, 34, 3523-3523 (1993)
M. Golinski et al.
The Journal of Organic Chemistry, 58, 159-159 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service