Skip to Content
Merck
All Photos(3)

Key Documents

432857

Sigma-Aldrich

Sodium tetrachloroplatinate(II) hydrate

Synonym(s):

Disodium Tetrachloroplatinate Hydrate, Platinum(II) sodium chloride, Sodium chloroplatinate(II)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€184.00
5 G
€732.00

€184.00


Estimated to ship on25 March 2025


Request a Bulk Order

Select a Size

Change View
1 G
€184.00
5 G
€732.00

About This Item

Linear Formula:
Na2PtCl4·xH2O
CAS Number:
Molecular Weight:
382.87 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid

€184.00


Estimated to ship on25 March 2025


Request a Bulk Order

form

solid

Quality Level

composition

Degree of hydration, ~1
Pt, 48%

concentration

≤49.8% Pt (gravimetric)

SMILES string

O.[Na+].[Na+].Cl[Pt--](Cl)(Cl)Cl

InChI

1S/4ClH.2Na.H2O.Pt/h4*1H;;;1H2;/q;;;;2*+1;;+2/p-4

InChI key

GIEPPPFPHULPBK-UHFFFAOYSA-J

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiwei Sun et al.
Advanced materials (Deerfield Beach, Fla.), 27(29), 4364-4370 (2015-06-20)
Low molecular weight P2VP-b-PS-b-P2VP triblock copolymer (poly(2-vinlypyridine)-block-polystyrene-block-poly(2-vinylpyridine)] is doped with copper chloride and microphase separated into lamellar line patterns with ultrahigh area density. Salt-doped P2VP-b-PS-b-P2VP triblock copolymer is self-assembled on the top of the nanoimprinted photoresist template, and metallic nanowires

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service