Skip to Content
Merck
All Photos(2)

Key Documents

432407

Sigma-Aldrich

Polystyrene-block-polyisoprene-block-polystyrene

styrene 17 wt. %

Synonym(s):

Polystyrene-block-polyisoprene, multi-arm

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€79.30

€79.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€79.30

About This Item

Linear Formula:
[CH2CH(C6H5)]X[CH2CH=C(CH3)CH2]Y[CH2CH(C6H5)]z
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€79.30


Check Cart for Availability

Request a Bulk Order

form

beads

Quality Level

melt index

106 g/10 min (200°C/5.0kg)

mol wt

Mn 1,900

contains

phenolic as antioxidant

composition

styrene, 17 wt. %

hardness

32 (Shore A)

viscosity

5 poise, 25 wt. % in toluene(25 °C, Brookfield)(lit.)

solubility

solvents with solubility parameters between 7.7 and 9.4: soluble

density

0.92 g/mL at 25 °C

elongation

ASTM D 412 - 1,300% (ultimate)

SMILES string

CC(=C)C=C.C=Cc1ccccc1

InChI

1S/C8H8.C5H8/c1-2-8-6-4-3-5-7-8;1-4-5(2)3/h2-7H,1H2;4H,1-2H2,3H3

InChI key

ROGIWVXWXZRRMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Component of hot melt adhesives, sealants, asphalt and oil gels. Modifier for polymers, thermosets and general rubber compounding. Direct molded or extruded auto parts, sporting goods, footware and film.

Features and Benefits

High strength and low viscosity for easy melt and solution processing. High temperature degradation results in cross-linking. Flexible to -65°C. Excellent electrical insulator, resists acids and alkali.

Physical form

Thermoplastic elastomer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph D Sandt et al.
Advanced healthcare materials, 7(15), e1800293-e1800293 (2018-05-29)
Medical textiles are widely used to exert pressure on human tissues during treatment of post-surgical hematoma, burn-related wounds, chronic venous ulceration, and other maladies. However, the inability to dynamically sense and adjust the applied pressure often leads to suboptimal pressure

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service